Novus Biologicals
Manufacturer Code:NBP107100JF646
Catalog # NBP17100646
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | J4B6 |
Immunogen | Recombinant human TMEFF2 (41-150aa) purified from E. coli |
Conjugate | Janelia Fluor 646 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: cancer/testis antigen family 120, member 2; CT120.2 HPP1member 2 Hyperplastic polyposis protein 1 TPEFtomoregulin TR-2 transmembrane protein TENB2 Transmembrane protein with EGF-like and two follistatin-like domains transmembrane protein with EGF-like and two follistatin-like domains 2 TRtomoregulin-2; Hyperplastic polyposis protein 1; tomoregulin; Tomoregulin-2; TR-2; transmembrane protein TENB2; Transmembrane protein with EGF-like and two follistatin-like domains; transmembrane protein with EGF-like and two follistatin-like domains 2
Gene Aliases: CT120.2; HPP1; TENB2; TMEFF2; TPEF; TR; TR-2; UNQ178/PRO204
UniProt ID: (Human) Q9UIK5
Entrez Gene ID: (Human) 23671
Molecular Function: extracellular matrix linker protein extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.