Novus Biologicals
Manufacturer Code:NBP131650
Catalog # NBP131650
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human TMED9. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glycoprotein 25L2; Glycoprotein 25L2 GP25L2 gp25L2 protein HSGP25L2G transmembrane emp24 domain-containing protein 9 transmembrane emp24 protein transport domain containing 9; GMP25; gp25L2 protein; p24 family protein alpha-2; p24alpha2; p25; Transmembrane emp24 domain-containing protein 9; transmembrane emp24 protein transport domain containing 9
Gene Aliases: GMP25; GP25L2; HSGP25L2G; p24a2; p25; TMED9
UniProt ID: (Human) Q9BVK6
Entrez Gene ID: (Human) 54732
Molecular Function: membrane traffic protein transfer/carrier protein vesicle coat protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.