Proteintech
Manufacturer Code:216201AP150UL
Catalog # 50-173-3516
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TMED9 Fusion Protein Ag13813 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glycoprotein 25L2; Glycoprotein 25L2 GMP25 GP25L2 HSGP25L2G TMED9; GMP25; gp25L2 protein; p24 family protein alpha-2; p24alpha2; p25; Transmembrane emp24 domain-containing protein 9; transmembrane emp24 protein transport domain containing 9
Gene Aliases: 2400003B06Rik; GMP25; GP25L2; HSGP25L2G; p24a2; p25; RGD1307627; TMED9
UniProt ID: (Human) Q9BVK6, (Rat) Q5I0E7, (Mouse) Q99KF1
Entrez Gene ID: (Human) 54732, (Rat) 361207, (Mouse) 67511
Molecular Function:
membrane traffic protein
transfer/carrier protein
vesicle coat protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.