Novus Biologicals
Manufacturer Code:NBP2836630.1ML
Catalog # NB031592
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human TMED2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: coated vesicle membrane protein; FLJ21323 Membrane protein p24A P24A RNP24coated vesicle membrane protein transmembrane emp24 domain trafficking protein 2 transmembrane emp24 domain-containing protein 2; Membrane protein p24A; p24; p24 family protein beta-1; p24beta1; transmembrane emp24 domain trafficking protein 2; Transmembrane emp24 domain-containing protein 2
Gene Aliases: p24; P24A; p24b1; p24beta1; RNP24; TMED2
UniProt ID: (Human) Q15363
Entrez Gene ID: (Human) 10959
Molecular Function: membrane traffic protein transfer/carrier protein vesicle coat protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.