Abnova
Manufacturer Code:PAB20694
Catalog # 89-122-196
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human TMED2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant TMED2.
For Research Use Only
Protein Aliases: coated vesicle membrane protein; FLJ21323; Membrane protein p24A; p24; p24 family protein beta-1; P24A; p24beta1; RNP24; transmembrane emp24 domain trafficking protein 2; Transmembrane emp24 domain-containing protein 2
Gene Aliases: p24; P24A; p24b1; p24beta1; RNP24; TMED2
UniProt ID: (Human) Q15363
Entrez Gene ID: (Human) 10959
Molecular Function:
membrane traffic protein
transfer/carrier protein
vesicle coat protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.