Invitrogen
Manufacturer Code:OSR00227W
Catalog # PIOSR00227W
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide from the 1t cytoplasmic loop of mouse transmembrane channel-like protein 2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beethoven transmembrane channel-like protein 2 transmembrane cochlear-expressed protein 2 dn Bth CWEA1 4933416G09Rik; Transmembrane channel-like protein 2; Transmembrane cochlear-expressed protein 2; transmembrane, cochlear expressed 2
Gene Aliases: CWEA2; Tmc2
UniProt ID: (Mouse) Q8R4P4
Entrez Gene ID: (Mouse) 192140
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.