Abnova
Manufacturer Code:PAB11278
Catalog # 89-114-985
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to N-terminus of human TMBIM1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against partial recombinant TMBIM1.
For Research Use Only
Protein Aliases: MST100; MSTP100; PP1201; Protein lifeguard 3; Protein RECS1 homolog; RECS1; Transmembrane BAX inhibitor motif-containing protein 1
Gene Aliases: LFG3; MST100; MSTP100; PP1201; PSEC0158; RECS1; TMBIM1
UniProt ID: (Human) Q969X1
Entrez Gene ID: (Human) 64114
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.