Invitrogen
Manufacturer Code:PA556540
Catalog # PIPA556540
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human TM7SF2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-beta-hydroxysterol Delta (14)-reductase; Another new gene 1 protein; C-14 sterol reductase; C14SR; Delta(14)-sterol reductase TM7SF2; Delta-14-SR; Putative sterol reductase SR-1; Sterol C14-reductase; Transmembrane 7 superfamily member 2
Gene Aliases: ANG1; DHCR14A; NET47; TM7SF2
UniProt ID: (Human) O76062
Entrez Gene ID: (Human) 7108
Molecular Function:
oxidoreductase
receptor
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.