Invitrogen
Manufacturer Code:PA5110492
Catalog # PIPA5110492
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 160-250 of human TM7SF2 (NP_003264.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-beta-hydroxysterol Delta (14)-reductase; 3110041O18Rik 3-beta-hydroxysterol Delta (14)-reductase ANG1 Another new gene 1 protein BOS_24949 C-14 sterol reductase C14SR Delta(14)-sterol reductase TM7SF2 Delta-14-SR Dhcr14 DHCR14A NET47 Putative sterol reductase SR-1 Sterol C14-reductase Tm7sf2 Transmembrane 7 superfamily member 2; Another new gene 1 protein; C-14 sterol reductase; C14SR; delta(14)-sterol reductase; Delta(14)-sterol reductase TM7SF2; Delta-14-SR; Putative sterol reductase SR-1; Sterol C14-reductase; Transmembrane 7 superfamily member 2
Gene Aliases: 3110041O18Rik; ANG1; C14SR; Dhcr14; DHCR14A; NET47; TM7SF2
UniProt ID: (Human) O76062, (Mouse) Q71KT5
Entrez Gene ID: (Human) 7108, (Mouse) 73166
Molecular Function:
oxidoreductase
receptor
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.