R&D Systems
Manufacturer Code:PPH650600
Catalog # PPH650600
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | H6506 |
Immunogen | Recombinant human TLX (amino acids 81 - 180) and mouse myeloma cells (NS-1) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at -70 degreesC. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: hTll NR2E1 nuclear receptor subfamily 2 group E member 1 Nuclear receptor TLX Protein tailless homolog tailes-related receptor tailless homolog TLL TLXnuclear receptor subfamily 2 group E member 1 XTLL; Nuclear receptor subfamily 2 group E member 1; nuclear receptor subfamily 2, group E, member 1; Nuclear receptor TLX; Protein tailless homolog; tailes-related receptor; Tll
Gene Aliases: NR2E1; TLL; TLX; XTLL
UniProt ID: (Human) Q9Y466
Entrez Gene ID: (Human) 7101
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.