Novus Biologicals
Manufacturer Code:NBP238764
Catalog # NBP238764
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: IPVDANTLLAVSGMNGDNTDSQKLNKIISEIQALQEVVARFRQLRLDATEFACLKCIVTFKAVPTHSGSELRSFRNAAAIAALQDEAQLTLNSY |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hTll nuclear receptor subfamily 2 group E member 1 Nuclear receptor TLX Protein tailless homolog tailes-related receptor tailless homolog Tll TLXnuclear receptor subfamily 2 group E member 1 XTLL; Nuclear receptor subfamily 2 group E member 1; nuclear receptor subfamily 2, group E, member 1; Nuclear receptor TLX; Protein tailless homolog; tailes-related receptor; Tll
Gene Aliases: NR2E1; TLL; TLX; XTLL
UniProt ID: (Human) Q9Y466
Entrez Gene ID: (Human) 7101
Molecular Function:
nuclear hormone receptor
nucleic acid binding
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.