Novus Biologicals
Manufacturer Code:NBP231150
Catalog # NBP231150
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 2A4C6.2E5 |
Immunogen | Partial recombinant protein made to an internal portion of mouse TLR9 (between amino acids 500-800) [UniProt Q9EQU3] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CD289; CD289 CD289 antigen toll-like receptor 9; Toll-like receptor 9
Gene Aliases: CD289; TLR9; UNQ5798/PRO19605
UniProt ID: (Human) Q9NR96
Entrez Gene ID: (Human) 54106
Molecular Function:
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.