Novus Biologicals
Manufacturer Code:NBP118284SS
Catalog # NBP118284SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide made to an internal portion of the human TLR9 protein (within residues 1019-1032). [Swiss-Prot# Q9NR96] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD289; CD289 CD289 antigen toll-like receptor 9; Toll-like receptor 9
Gene Aliases: CD289; TLR9; UNQ5798/PRO19605
UniProt ID: (Human) Q9NR96
Entrez Gene ID: (Human) 54106
Molecular Function: extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.