Novus Biologicals
Manufacturer Code:NBP224969
Catalog # NBP224969
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 86B1153.2 |
Immunogen | This antibody was developed against a synthetic peptide corresponding to amino acids 408-424 of human TRL6. |
Conjugate | PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD286; CD286 CD286 antigen toll-like receptor 6; Toll-like receptor 6
Gene Aliases: CD286; TLR6
UniProt ID: (Human) Q9Y2C9
Entrez Gene ID: (Human) 10333
Molecular Function:
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.