Novus Biologicals
Manufacturer Code:NBP224787
Catalog # NBP224787
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 19D759.2 |
Immunogen | This antibody was developed against KLH-conjugated synthetic peptide corresponding to a portion of human TLR5 between amino acids 700-800. The antibody also reacts with mouse TLR5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: MGC126430 MGC126431 SLEB1FLJ10052 TIL3Toll/interleukin-1 receptor-like protein 3 toll-like receptor 5; systemic lupus erythematosus susceptibility 1; Toll-like receptor 5; Toll/interleukin-1 receptor-like protein 3
Gene Aliases: MELIOS; SLE1; SLEB1; TIL3; TLR5
UniProt ID: (Human) O60602
Entrez Gene ID: (Human) 7100
Molecular Function: extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.