Invitrogen
Manufacturer Code:25992482
Catalog # 50-156-26
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | MTS510 |
Conjugate | PE-Cyanine7 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CD284; ESOP-1; lipopolysaccharide response; Ly-96; Lymphocyte antigen 96; myeloid differentiation factor-2; myeloid differentiation protein-2; Protein MD-2; Toll-like receptor 4
Gene Aliases: ESOP-1; Esop1; Lps; Ly87; Ly96; MD-2; MD2; Ran/M1; Rasl2-8; Tlr4
UniProt ID: (Mouse) Q9JHF9, (Mouse) Q9QUK6
Entrez Gene ID: (Mouse) 17087, (Mouse) 21898
Molecular Function:
transmembrane receptor regulatory/adaptor protein
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.