Invitrogen
Manufacturer Code:MA516209
Catalog # PIMA516209
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | HTA125 |
Immunogen | Ba/F3 cell line expressing human TLR4 cell surface antigen |
Conjugate | PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD284; homolog of Drosophila toll; hToll; lipopolysaccharide response; Toll-like receptor 4; Toll-like Receptor 4 ARMD10 TLR-4 TOLL hToll homolog of Drosophila toll; Toll-like receptor4 protein
Gene Aliases: ARMD10; CD284; Lps; Ly87; Ran/M1; Rasl2-8; TLR-4; TLR4; TOLL
UniProt ID: (Human) O00206, (Mouse) Q9QUK6
Entrez Gene ID: (Pig) 399541, (Human) 7099, (Dog) 403417, (Mouse) 21898, (Guinea pig) 100724803
Molecular Function: extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.