Novus Biologicals
Manufacturer Code:NB10056727
Catalog # NB10056727
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunoblot (IB) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
In vitro Assay (IV) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | HTA125 |
Immunogen | This antibody was developed by immunizing mice with Ba/F3 cell line expressing human TLR4 cell surface antigen. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ARMD10 CD284 CD284 antigen hTollhomolog of Drosophila toll TOLL toll-like receptor 4; CD284; homolog of Drosophila toll; hToll; Toll-like receptor 4
Gene Aliases: ARMD10; CD284; TLR-4; TLR4; TOLL
UniProt ID: (Human) O00206
Entrez Gene ID: (Human) 7099
Molecular Function:
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.