Invitrogen
Manufacturer Code:710185
Catalog # PI710185
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Oligoclonal |
Type | Antibody |
Clone | 20HCLC |
Immunogen | Peptide corresponding to amino acids 816-830 of human Toll-like receptor 4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Polyclonal Antibody
For Research Use Only
Protein Aliases: CD284; homolog of Drosophila toll; hToll; lipopolysaccharide response; Toll-like receptor 4; Toll-like Receptor 4 ARMD10 CD284 TLR-4 TOLL hToll homolog of Drosophila toll
Gene Aliases: ARMD10; CD284; Lps; Ly87; Ran/M1; Rasl2-8; TLR-4; TLR4; TOLL
UniProt ID: (Human) O00206, (Mouse) Q9QUK6
Entrez Gene ID: (Human) 7099, (Mouse) 21898
Molecular Function: extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.