Invitrogen
Manufacturer Code:PA511597
Catalog # PIPA511597
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 25-55 amino acids from the N-terminal region of mouse TLR4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD284; lipopolysaccharide response; Toll-like receptor 4; Toll-like Receptor 4 ARMD10 TLR-4 TOLL hToll homolog of Drosophila toll
Gene Aliases: Lps; Ly87; Ran/M1; Rasl2-8; Tlr4
UniProt ID: (Mouse) Q9QUK6
Entrez Gene ID: (Mouse) 21898
Molecular Function:
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.