Novus Biologicals
Manufacturer Code:NB10056571
Catalog # NB10056571
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A mix of synthetic peptides corresponding to amino acids 135-150 (SIHKIKSNPFKNQKNL) 828-844 (CRRFKVHHAVQQAIEQN) and 876-891 (CILNWPVQKERINAFH) of mouse TLR3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD283; CD283 CD283 antigen toll-like receptor 3; Toll-like receptor 3
Gene Aliases: CD283; IIAE2; TLR3
UniProt ID: (Human) O15455
Entrez Gene ID: (Human) 7098
Molecular Function:
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.