Proteintech
Manufacturer Code:177661AP150UL
Catalog # 50-173-1777
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TLR3 Fusion Protein Ag12171 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD283; CD283 TLR3 toll like receptor 3; Toll-like receptor 3
Gene Aliases: AI957183; CD283; IIAE2; TLR3
UniProt ID: (Human) O15455, (Mouse) Q99MB1
Entrez Gene ID: (Human) 7098, (Mouse) 142980, (Rat) 364594
Molecular Function: extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.