Novus Biologicals
Manufacturer Code:NBP177229
Catalog # NBP177229
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a peptide corresponding to 14 amino acids near the amino terminus of human TLR2. The immunogen is located within the first 50 amino acids of TLR2. Amino Acid Squence: CKGSSGSLNSIPSG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD282; CD282 antigen TIL4CD282 toll/interleukin 1 receptor-like 4 Toll/interleukin-1 receptor-like protein 4 toll-like receptor 2; Toll-like receptor 2; Toll/interleukin-1 receptor-like protein 4
Gene Aliases: CD282; TIL4; TLR2
UniProt ID: (Human) O60603
Entrez Gene ID: (Human) 7097
Molecular Function: extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.