Invitrogen
Manufacturer Code:PA141480
Catalog # 01-675-308
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to residues 180-196 353-370 and 473-489 of human TLR2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD282; Toll-like receptor 2; Toll-like Receptor 2 TIL4 toll/interleukin 1 receptor-like 4 toll/interleukin-1 receptor-like protein 4; Toll/interleukin-1 receptor-like protein 4
Gene Aliases: CD282; Ly105; TIL4; TLR2
UniProt ID: (Human) O60603, (Mouse) Q9QUN7
Entrez Gene ID: (Human) 7097, (Mouse) 24088
Molecular Function:
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.