Novus Biologicals
Manufacturer Code:NB10056563
Catalog # NB10056563
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Dot blot (DB) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a KLH-conjugated synthetic peptide corresponding to a sequence within amino acids 400-450 of human TLR1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD281; CD281 CD281 antigen DKFZp547I0610 DKFZp564I0682 KIAA0012MGC126311 MGC104956 MGC126312 rsc786 TIL TIL. LPRS5 Toll/interleukin-1 receptor-like protein toll-like receptor 1; TIL; Toll-like receptor 1; Toll/interleukin-1 receptor-like protein
Gene Aliases: CD281; KIAA0012; rsc786; TIL; TIL. LPRS5; TLR1
UniProt ID: (Human) Q15399
Entrez Gene ID: (Human) 7096
Molecular Function:
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.