R&D Systems
Manufacturer Code:FAB1484P
Catalog # FAB1484P
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human TLR1 Ser22-Asn578 Accession # AAC34137 |
Conjugate | R-PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
For Research Use Only
Protein Aliases: CD281; CD281 CD281 antigen DKFZp547I0610 DKFZp564I0682 KIAA0012MGC126311 MGC104956 MGC126312 rsc786 TIL TIL. LPRS5 Toll/interleukin-1 receptor-like protein toll-like receptor 1; TIL; Toll-like receptor 1; Toll/interleukin-1 receptor-like protein
Gene Aliases: CD281; KIAA0012; rsc786; TIL; TIL. LPRS5; TLR1
UniProt ID: (Human) Q15399
Entrez Gene ID: (Human) 7096
Molecular Function: extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.