OriGene
Manufacturer Code:TA805492
Catalog # 50-168-0725
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2H2 |
Immunogen | Human recombit protein fragment corresponding to amino acids 1-296 of human TLK2 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: HsHPK; HsHPK PKU-ALPHA; PKU-alpha; Serine/threonine-protein kinase tousled-like 2; Tousled-like kinase 2
Gene Aliases: HsHPK; PKU-ALPHA; TLK2
UniProt ID: (Human) Q86UE8
Entrez Gene ID: (Human) 11011
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.