Invitrogen
Manufacturer Code:PA577019
Catalog # PIPA577019
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human TLK2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HsHPK; PKU-alpha; protein kinase U-alpha; Serine/threonine-protein kinase tousled-like 2; Serine/threonine-protein kinase tousled-like 2 HsHPK PKU-alpha Tousled-like kinase 2 TLK2; Tousled-like kinase 2
Gene Aliases: 4933403M19Rik; HsHPK; PKU-ALPHA; PKUalpha; Tlk; TLK2
UniProt ID: (Human) Q86UE8, (Mouse) O55047
Entrez Gene ID: (Human) 11011, (Mouse) 24086
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.