Invitrogen
Manufacturer Code:PA515375
Catalog # PIPA515375
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 141-171 amino acids from the central region of human TLK2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HsHPK; PKU-alpha; protein kinase U-alpha; Serine/threonine-protein kinase tousled-like 2; Serine/threonine-protein kinase tousled-like 2 Tousled-like kinase 2 PKU-alpha; Tousled-like kinase 2
Gene Aliases: 4933403M19Rik; HsHPK; PKU-ALPHA; PKUalpha; Tlk; TLK2
UniProt ID: (Human) Q86UE8, (Mouse) O55047
Entrez Gene ID: (Human) 11011, (Mouse) 24086
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.