Novus Biologicals
Manufacturer Code:NBP15435520UL
Catalog # NBP15435520
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to TLK2(tousled-like kinase 2) The peptide sequence was selected from the N terminal of TLK2. Peptide sequence VSAQQNSPSSTGSGNTEHSCSSQKQISIQHRQTQSDLTIEKISALENSKN. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.11 EC 2.7.11.1 MGC44450 PKU-ALPHA serine/threonine-protein kinase tousled-like 2 tousled-like kinase 2serine/threonine kinase; HsHPK; PKU-alpha; Serine/threonine-protein kinase tousled-like 2; Tousled-like kinase 2
Gene Aliases: HsHPK; PKU-ALPHA; TLK2
UniProt ID: (Human) Q86UE8
Entrez Gene ID: (Human) 11011
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.