Proteintech
Manufacturer Code:139791AP150UL
Catalog # 50-172-9136
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TLK2 Fusion Protein Ag5022 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HsHPK; PKU ALPHA TLK2 tousled like kinase 2; PKU-alpha; protein kinase U-alpha; Serine/threonine-protein kinase tousled-like 2; Tousled-like kinase 2
Gene Aliases: 4933403M19Rik; HsHPK; PKU-ALPHA; PKUalpha; Tlk; TLK2
UniProt ID: (Human) Q86UE8, (Mouse) O55047
Entrez Gene ID: (Human) 11011, (Rat) 303592, (Mouse) 24086
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.