Novus Biologicals
Manufacturer Code:NBP246134
Catalog # NBP246134
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1H2 |
Immunogen | Human recombinant protein fragment corresponding to amino acids 180-460 of human TLE1 (NP_005068) produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: E(Sp1) homolog; E(Sp1) homolog enhancer of split groucho 1 ESG ESG1Enhancer of split groucho-like protein 1 GRG1 transducin-like enhancer of split 1 (E(sp1) homolog Drosophila) transducin-like enhancer of split 1 homolog of Drosophila E(sp1) transducin-like enhancer protein 1; Enhancer of split groucho-like protein 1; ESG1; transducin-like enhancer of split 1 (E(sp1) homolog, Drosophila); Transducin-like enhancer protein 1
Gene Aliases: ESG; ESG1; GRG1; TLE1
UniProt ID: (Human) Q04724
Entrez Gene ID: (Human) 7088
Molecular Function:
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.