Novus Biologicals
Manufacturer Code:NBP281120AF594
Catalog # NB016558
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | TAN2-2 |
Immunogen | This antibody was raised by immunising rats with a soluble recombinant TL1A/TNFSF15 (T77-L252):Fc protein. |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: MGC129934 MGC129935 TL1A TL1Vascular endothelial cell growth inhibitor TNF superfamily ligand TL1A tumor necrosis factor (ligand) superfamily member 15 tumor necrosis factor ligand superfamily member 15 vascular endothelial growth inhibitor-192A VEGI192A VEGITNF ligand-related molecule 1; TNF ligand-related molecule 1; TNF superfamily ligand TL1A; tumor necrosis factor (ligand) superfamily member 15; tumor necrosis factor (ligand) superfamily, member 15; tumor necrosis factor ligand 1B; Tumor necrosis factor ligand superfamily member 15; Tumor necrosis factor ligand superfamily member 15, membrane form; Tumor necrosis factor ligand superfamily member 15, secreted form; Vascular endothelial cell growth inhibitor; vascular endothelial growth inhibitor-192A
Gene Aliases: TL1; TL1A; TNFSF15; TNLG1B; VEGI; VEGI192A
UniProt ID: (Human) O95150
Entrez Gene ID: (Human) 9966
Molecular Function:
cytokine
signaling molecule
tumor necrosis factor family member
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.