R&D Systems
Manufacturer Code:MAB18961100
Catalog # MAB18961100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 820422 |
Immunogen | <em>E. coli</em>-derived recombinant mouse TL1A/TNFSF15 Ile76-Leu252 Accession # Q5UBV8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70°C as supplied. 1 month 2 to 8°C under sterile conditions after reconstitution. 6 months -20 to -70°C under sterile conditions a |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: MGC129934 MGC129935 TL1A TL1Vascular endothelial cell growth inhibitor TNF superfamily ligand TL1A TNFSF15 tumor necrosis factor (ligand) superfamily member 15 tumor necrosis factor ligand superfamily member 15 vascular endothelial growth inhibitor-192A VEGI VEGI192A VEGITNF ligand-related molecule 1; TNF ligand-related molecule 1; TNF superfamily ligand TL1A; tumor necrosis factor (ligand) superfamily member 15; tumor necrosis factor (ligand) superfamily, member 15; tumor necrosis factor ligand 1B; Tumor necrosis factor ligand superfamily member 15; Tumor necrosis factor ligand superfamily member 15, membrane form; Tumor necrosis factor ligand superfamily member 15, secreted form; Vascular endothelial cell growth inhibitor; vascular endothelial growth inhibitor-192A
Gene Aliases: TL1; TL1A; TNFSF15; TNLG1B; VEGI; VEGI192A
UniProt ID: (Human) O95150
Entrez Gene ID: (Human) 9966
Molecular Function: cytokine signaling molecule tumor necrosis factor family member
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.