Invitrogen
Manufacturer Code:PA531193
Catalog # PIPA531193
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 244 of Human TKTL2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: testis tissue sperm-binding protein Li 39a; transketolase-like 2; Transketolase-like protein 2
Gene Aliases: 4933401I19Rik; TKTL2
UniProt ID: (Human) Q9H0I9, (Mouse) Q9D4D4
Entrez Gene ID: (Human) 84076, (Mouse) 74419
Molecular Function: dehydrogenase lyase oxidoreductase transferase transketolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.