Invitrogen
Manufacturer Code:PA544093
Catalog # PIPA544093
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human TKTL1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: TK 2; tktl1; Transketolase 2; transketolase-2; transketolase-like 1; Transketolase-like 1 TKR TKT2; Transketolase-like protein 1; Transketolase-related protein
Gene Aliases: C79967; TKR; TKT2; TKTL1
UniProt ID: (Human) P51854, (Mouse) Q99MX0
Entrez Gene ID: (Human) 8277, (Horse) 100059329, (Rabbit) 100354239, (Dog) 481083, (Guinea pig) 100725793, (Mouse) 83553, (Rat) 689374
Molecular Function:
dehydrogenase
lyase
oxidoreductase
transferase
transketolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.