Invitrogen
Manufacturer Code:PA528977
Catalog # PIPA528977
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 311 of Human TKTL1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: TK 2; TKR TKT2 TK 2 transketolase-2 transketolase-like protein 1 transketolase-related protein; Transketolase 2; transketolase-2; transketolase-like 1; Transketolase-like protein 1; Transketolase-related protein
Gene Aliases: TKR; TKT2; TKTL1
UniProt ID: (Human) P51854
Entrez Gene ID: (Human) 8277
Molecular Function:
dehydrogenase
lyase
oxidoreductase
transferase
transketolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.