Abnova
Manufacturer Code:H00009414K
Catalog # 89-203-239
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human TJP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human TJP2 peptide using ARM Technology.
For Research Use Only
Protein Aliases: Friedreich ataxia region gene X104 (tight junction protein ZO-2); MGC26306 X104 ZO-2 ZO2; Tight junction protein 2; Tight junction protein ZO-2; zona occludens 2; Zona occludens protein 2; Zonula occludens protein 2
Gene Aliases: C9DUPq21.11; DFNA51; DUP9q21.11; PFIC4; TJP2; X104; ZO2
UniProt ID: (Human) Q9UDY2
Entrez Gene ID: (Human) 9414
Molecular Function:
cell junction protein
tight junction
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.