Invitrogen
Manufacturer Code:PA520028
Catalog # PIPA520028
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A peptide corresponding to 15 amino acids near the C-terminus of mouse TIRAP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adapter protein wyatt; Adaptor protein Wyatt; MyD88 adapter-like protein; MyD88-2; TIR domain-containing adapter protein; toll-interleukin 1 receptor (TIR) domain containing adaptor protein; toll-interleukin 1 receptor (TIR) domain-containing adaptor protein; Toll-like receptor 4 adaptor protein; Toll-like receptor adaptor protein; Toll/interleukin-1 receptor domain-containing adapter protein
Gene Aliases: AA407980; BACTS1; C130027E04Rik; MAL; MyD88-2; TIRAP; Tlr4ap; Wyatt
UniProt ID: (Human) P58753, (Mouse) Q99JY1
Entrez Gene ID: (Human) 114609, (Mouse) 117149, (Rat) 680127
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.