Abnova
Manufacturer Code:PAB9785
Catalog # 89-134-414
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus of mouse Tirap. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Tirap.
For Research Use Only
Protein Aliases: AA407980; Adaptor protein Wyatt; C130027E04Rik; Mal; MyD88 adapter-like protein; TIR domain-containing adapter protein; Tlr4ap; Toll-like receptor 4 adaptor protein; Toll/interleukin-1 receptor domain-containing adapter protein; wyatt
Gene Aliases: AA407980; C130027E04Rik; Mal; Tirap; Tlr4ap; Wyatt
UniProt ID: (Mouse) Q99JY1
Entrez Gene ID: (Mouse) 117149
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.