Novus Biologicals
Manufacturer Code:NBP246133
Catalog # NBP246133
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2E6 |
Immunogen | Full length human recombinant protein of human TIPIN (NP_060328) produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CSM3 homolog; FLJ20516 TIMELESS interacting protein TIMELESS-interacting protein; TIMELESS-interacting protein
Gene Aliases: TIPIN
UniProt ID: (Human) Q9BVW5
Entrez Gene ID: (Human) 54962
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.