Invitrogen
Manufacturer Code:PA540774
Catalog # PIPA540774
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human TIPARP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADP-ribosyltransferase diphtheria toxin-like 14; ARTD14; PARP-7; Poly [ADP-ribose] polymerase 7; Protein mono-ADP-ribosyltransferase TIPARP; TCDD-inducible poly [ADP-ribose] polymerase; TCDD-inducible poly(ADP-ribose) polymerase; TCDD-inducible poly(ADP-ribose) polymerase DDF1 DKFZP434J214 DKFZp686N0351 DKFZp686P1838 FLJ40466 PARP-1 PARP-7 PARP7 pART14
Gene Aliases: ARTD14; AW558171; PARP7; pART14; RM1; TIPARP
UniProt ID: (Human) Q86VP4, (Mouse) Q8C1B2
Entrez Gene ID: (Rabbit) 100340932, (Dog) 485728, (Human) 25976, (Horse) 100050358, (Bovine) 540975, (Pig) 100626111, (Rat) 310467, (Mouse) 99929
Molecular Function: nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.