Abnova
Manufacturer Code:PAB23059
Catalog # 89-124-758
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human TIPARP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant TIPARP.
For Research Use Only
Protein Aliases: ADP-ribosyltransferase diphtheria toxin-like 14; ARTD14; DDF1; DKFZp434J214; DKFZp686N0351; DKFZp686P1838; FLJ40466; PARP-1; PARP-7; PARP7; Poly [ADP-ribose] polymerase 7; Protein mono-ADP-ribosyltransferase TIPARP; TCDD-inducible poly [ADP-ribose] polymerase; TCDD-inducible poly(ADP-ribose) polymerase
Gene Aliases: ARTD14; PARP7; pART14; TIPARP
UniProt ID: (Human) Q86VP4
Entrez Gene ID: (Human) 25976
Molecular Function: nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.