Novus Biologicals
Manufacturer Code:NBP157765
Catalog # NBP157765
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to TINAGL1 (tubulointerstitial nephritis antigen-like 1) The peptide sequence was selected from the middle region of TINAGL1)(50ug). Peptide sequence NLIHEPLDQGNCAGSWAFSTAAVASDRVSIHSLGHMTPVLSPQNLLSCDT. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: androgen-regulated gene 1; androgen-regulated gene 1 ARG1 GIS5 Glucocorticoid-inducible protein 5 LCN7 LIECG3 lipocalin 7 OLRG2 OLRG-2 Oxidized LDL-responsive gene 2 protein oxidized-LDL responsive gene 2 P3ECSL TIN Ag-related protein TINAGL TINAG-like 1 TIN-Ag-RP TINAGRP tubulointerstitial nephritis antigen-like tubulointerstitial nephritis antigen-like 1 Tubulointerstitial nephritis antigen-related protein; Glucocorticoid-inducible protein 5; lipocalin 7; OLRG-2; Oxidized LDL-responsive gene 2 protein; oxidized-LDL responsive gene 2; P3ECSL; TIN Ag-related protein; TIN-Ag-RP; TINAG-like 1; Tubulointerstitial nephritis antigen-like; tubulointerstitial nephritis antigen-like 1; Tubulointerstitial nephritis antigen-related protein
Gene Aliases: ARG1; GIS5; LCN7; LIECG3; OLRG2; PP6614; PSEC0088; TINAGL; TINAGL1; TINAGRP; UNQ204/PRO230
UniProt ID: (Human) Q9GZM7
Entrez Gene ID: (Human) 64129
Molecular Function: cysteine protease hydrolase protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.