Proteintech
Manufacturer Code:120771AP150UL
Catalog # 50-172-7695
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TINAGL1 Fusion Protein Ag2721 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adrenocortical zonation factor 1; androgen-regulated gene 1; Androgen-regulated gene 1 protein; ARG1 GIS5 LCN7 LIECG3 OLRG 2 OLRG2 PSEC0088 TIN Ag related protein TIN Ag RP TINAGL TINAGL1 TINAGRP UNQ204/PRO230; AZ-1; Glucocorticoid-inducible protein 5; lipocalin 7; OLRG-2; Oxidized LDL-responsive gene 2 protein; oxidized-LDL responsive gene 2; P3ECSL; TARP; TIN Ag-related protein; TIN-Ag-RP; TINAG-like 1; Tubulointerstitial nephritis antigen-like; tubulointerstitial nephritis antigen-like 1; Tubulointerstitial nephritis antigen-related protein
Gene Aliases: 1110021J17Rik; ARG1; AZ-1; AZ1; GIS5; LCN7; LIECG3; OLRG2; PP6614; PSEC0088; TARP; TINAGL; TINAGL1; TINAGRP; UNQ204/PRO230
UniProt ID: (Human) Q9GZM7, (Mouse) Q99JR5, (Rat) Q9EQT5
Entrez Gene ID: (Human) 64129, (Mouse) 94242, (Rat) 94174
Molecular Function: cysteine protease hydrolase protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.