Proteintech
Manufacturer Code:114791AP150UL
Catalog # 50-172-7196
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TIMM9 Fusion Protein Ag2050 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Mitochondrial import inner membrane translocase subunit Tim9; TIM9 TIM9A TIMM9 TIMM9A; translocase of inner mitochondrial membrane 10 homolog; translocase of inner mitochondrial membrane 9 homolog
Gene Aliases: 2810011L15Rik; Tim10a; TIM9; TIM9A; Timm10; TIMM9; TIMM9A
UniProt ID: (Human) Q9Y5J7, (Mouse) Q9WV98, (Rat) Q9WV97
Entrez Gene ID: (Human) 26520, (Mouse) 30056, (Rat) 171139
Molecular Function: mitochondrial carrier protein transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.