Novus Biologicals
Manufacturer Code:NBP294190100UL
Catalog # NB022937
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 16-98 of human TIMM8B (NP_036591.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DDP-like protein; DDP2DDP-like protein DDPL Deafness dystonia protein 2 FLJ21744 MGC102866 MGC117373 mitochondrial import inner membrane translocase subunit Tim8 B TIM8Btranslocase of inner mitochondrial membrane 8 (yeast) homolog B translocase of inner mitochondrial membrane 8 homolog B (yeast); Deafness dystonia protein 2; Mitochondrial import inner membrane translocase subunit Tim8 B
Gene Aliases: DDP2; DDPL; TIM8B; TIMM8B
UniProt ID: (Human) Q9Y5J9
Entrez Gene ID: (Human) 26521
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.