Abnova
Manufacturer Code:H00001678M01
Catalog # 89-021-295
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2F11 |
Immunogen | TIMM8A (NP_004076 9 a.a. ~ 97 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant TIMM8A.
For Research Use Only
Protein Aliases: DDP; DDP1; Deafness dystonia protein 1; deafness/dystonia peptide; DFN1; MGC12262; Mitochondrial import inner membrane translocase subunit Tim8 A; MTS; X-linked deafness dystonia protein
Gene Aliases: DDP; DDP1; DFN1; MTS; TIM8; TIM8A; TIMM8A
UniProt ID: (Human) O60220
Entrez Gene ID: (Human) 1678
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.