Invitrogen
Manufacturer Code:PA563305
Catalog # PIPA563305
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human TIM50 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: homolog of yeast Tim50; Mitochondrial import inner membrane translocase subunit TIM50; Tim50-like protein; TIMM50 TIM50L translocase of inner mitochondrial membrane 50 homolog (yeast) Tim50-like protein homolog of yeast Tim50 translocase of inner mitochondrial membrane 50 homolog (S. cerevisiae) MGC102733 TIM50; translocase of inner mitochondrial membrane 50 homolog
Gene Aliases: PRO1512; TIM50; TIM50L; TIMM50
UniProt ID: (Human) Q3ZCQ8
Entrez Gene ID: (Human) 92609
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.