Novus Biologicals
Manufacturer Code:NBP238791
Catalog # NBP238791
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: TGWRFKKRPGIETLFQQLAPLYEIVIFTSETGMTAFPLIDSVDPHGFISYRLFRDATRYMDGHHVKDISCLNRDPARVVVVDC |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: homolog of yeast Tim50; homolog of yeast Tim50 MGC102733 mitochondrial import inner membrane translocase subunit TIM50 TIM50 TIM50L Tim50-like protein translocase of inner mitochondrial membrane 50 homolog (S. cerevisiae) translocase of inner mitochondrial membrane 50 homolog (yeast); Mitochondrial import inner membrane translocase subunit TIM50; Tim50-like protein; translocase of inner mitochondrial membrane 50 homolog
Gene Aliases: PRO1512; TIM50; TIM50L; TIMM50
UniProt ID: (Human) Q3ZCQ8
Entrez Gene ID: (Human) 92609
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.